Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ADP Colorimetric Assay Kit II, BioVision

Supplier: BioVision
AB282932-100TEST
10186-656EA 1036.08 USD InStock
10186-656
ADP Colorimetric Assay Kit II, BioVision
Assays Cellular Assays
Highly Stable, HTS

Detection method - Absorbance (450 nm)
Application - The kit can detect less than 20 μM of ADP in samples.

ADP is a product of ATP dephosphorylation and it can be rephosphorylated to ATP. Dephosphorylation and rephosphorylation occur via various phosphorylases and kinases. ADP is stored in platelets and can be released to interact with variety of purinergic receptors. ADP levels regulate several enzymes involved in intermediary metabolism. ADP conversion to ATP primarily occurs within the mitochondrion and chloroplast although several such processes occur in the cytoplasm. Conventionally, ADP levels are measured by luciferase/luciferin mediated assays after ADP is converted to ATP. However, the luciferase system is unstable and luminescence equipment is not generally available in most laboratories. BioVision ADP Assay kit II is suitable for measuring ADP levels in samples that contain reducing substances, which may interfere with oxidase-based assays. In this assay, ADP in the presence of ADP Enzyme Mix is converted to an intermediate, which reduces a colorless Probe to a colored product with strong absorbance at 450 nm. ADP Assay Kit II is simple, fast and high-throughput ready. It can detect less than 20 μM of ADP in samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty