Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

FAD Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB204710-100TEST
10186-658EA 812.11 USD InStock
10186-658
FAD Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Sensitive Assay, HTS

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- FAD functions as the cofactor of an oxidase which catalyze the formation of a product that reacts with OxiRed probe generating color and fluorescence.The lower limit of detection is less than 1 nM FAD.

Flavin Adenine Dinucleotide (FAD) is a redox cofactor which plays an important role in metabolism. FAD exists in different redox states and cycles between FAD, FADH and FADH2. The primary sources of reduced FAD in eukaryotic metabolism are the citric acid cycle and the beta oxidation reaction pathways. In BioVision’s FAD Assay Kit, FAD functions as the cofactor of an oxidase which catalyze the formation of a product that reacts with OxiRed probe generating color and fluorescence. FAD can be detected by either colorimetric (λ=570 nm) or fluorometric (Ex/Em=535/587 nm) methods. The kit provides a rapid, simple, ultra-sensitive, and reliable test suitable for high throughput assay of FAD. The lower limit of detection is less than 1 nM FAD.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty