Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TACE Inhibitor Screening Assay Kit ( Fluorometric), BioVision

Supplier: BioVision
AB155889-100TEST
10186-662EA 1011.23 USD InStock
10186-662
TACE Inhibitor Screening Assay Kit ( Fluorometric), BioVision
Assays Cellular Assays
Sensitive Assay, HTS

Detection method- Fluorescence (Ex/Em 318/449 nm)
Species reactivity- Mammalian
Applications- Provides a rapid, simple, sensitive and reliable test suitable for high-throughput screening of TACE inhibitors and can be modified to check the relative TACE activity. Inhibitor control GM6001 is included to compare the efficiency of test inhibitors.

The TACE (tumor necrosis factor-α-converting enzyme), also called ADAM metallopeptidase domain 17 (ADAM17), is a 70-kDa enzyme that belongs to the ADAM protein family of disintegrins and metalloproteases. TACE is believed to be involved in the processing of tumor necrosis factor alpha (TNF-α) at the surface of the cell, and from within the intracellular membranes of the trans-Golgi network. This process, which is also known as 'shedding', involves the cleavage and release of a soluble ectodomain from membrane-bound pro-proteins (such as pro-TNF-α), and is of known physiological importance. In BioVision’s TACE inhibitor screening Kit, TACE hydrolyzes the specific FRET substrate to release the quenched fluorescent group, which can be detected fluorometrically at Ex/Em = 318/449 nm. In the presence of the potent TACE inhibitor, the hydrolyzation of substrate will be impeded. The kit provides a rapid, simple, sensitive and reliable test suitable for high-throughput screening of TACE inhibitors and can be modified to check the relative TACE activity. Inhibitor Control GM6001 is included to compare the efficacy of test inhibitors.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty