Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

cGMP Direct Immunoassay Kit (Colorimetric), BioVision

Supplier: BioVision
AB65356-100
10186-670EA 557.4 USD InStock
10186-670
cGMP Direct Immunoassay Kit (Colorimetric), BioVision
Assays Cellular Assays
Highly Sensitive Assay

Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Applications- The kit can detect 0.04 -10 pmol/well (0.008 - 2 µM) cGMP samples.

Adenosine and guanosine 3’,5’-cyclic monophosphate (cAMP and cGMP) are important “second messengers” involved in many physiological processes. BioVision’s cGMP Direct Immunoassay Kit provides a direct competitive immunoassay for sensitive and quantitative determination of cGMP level in biological samples. The kit utilizes the recombinant Protein G coated plate to anchor cGMP polyclonal antibody. cGMP-HRP conjugates directly competes with cGMP from samples for binding to the cGMP specific antibody on the plate. After incubation and washing, the amount of cGMP-HRP bound to plate can easily be determined by reading OD450 nm. The intensity of OD450 nm is inversely proportional to the concentration of cGMP in samples. The kit provides a new acetylation procedure that improves detection signal significantly. The kit can detect 0.04 -10 pmol/well (0.008 - 2 µM) cGMP samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty