Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Urea Colorimetric Assay Kit II, BioVision

Supplier: BioVision
AB234052-100TEST
10186-674EA 984.27 USD InStock
10186-674
Urea Colorimetric Assay Kit II, BioVision
Assays Cellular Assays
Highly Sensitive Assay

Detection method- Absorbance (505 nm)
Applications:
- Measurement of urea level in various tissues/cells
- Analysis of liver and kidney function

Urea is a waste product which is produced in the liver, dissolved and transported in the blood (in a concentration of 1.6-7.5 mM), and excreted by the kidney. Urea plays a very important role in protein catabolism, removal of toxic ammonia from the body, and the countercurrent system which allows for reabsorption of water and critical ions in the nephrons. Urea concentration is an important indicator for the medical clinicians to assess function of the kidney and other organs in patients. BioVision’s Urea Assay Kit II is based on Jung’s method with a modification that delivers more robust and sensitive data. The urea is condensed with o-phthalaldehyde (OPA), followed by a reaction to form a colored product with strong absorbance at 505 nm. This assay kit is fast, sensitive & easy to use. It can measure less than 10 μM urea in 96-well plate assay.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty