Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB102505-250TEST
10186-676EA 880.62 USD InStock
10186-676
Calcium Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (575 nm)
Species reactivity- Mammalian
Applications- The kit can detect calcium concentration 0.4-100 mg/dL (0.1-25 mM).

Calcium is essential for all living organisms, where Ca²ᶧ sequestration and release into and out of the cytoplasm functions as a signal for many cellular processes. 99% of calcium is found in bones and teeth with the remaining 1% found in the blood and soft tissue. Serum calcium levels are tightly controlled (8.4-11.4 mg/dL) and any variation outside this range can have serious effects. Calcium plays a role in mediating the constriction and relaxation of blood vessels, nerve impulse transmission, muscle contraction, and hormone secretion. Calcium ion channels control the migration of calcium ions across cell membranes, permitting the activation and inhibition of a wide variety of enzymes. Causes of low calcium levels include chronic kidney failure, vitamin D deficiency, and low blood magnesium levels that can occur in severe alcoholism. BioVision’s Colorimetric Calcium Assay Kit utilizes the chromogenic complex (λ = 575 nm) formed between calcium ions and 0-cresolphthalein to provide a simple assay in the physiologically important range of calcium concentration 0.4-100 mg/dL (0.1-25 mM).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty