Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB102506-100TEST
10186-678EA 877.69 USD InStock
10186-678
Magnesium Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Applications- The linear range of the assay is 2-15 nmoles with detection sensitivity~ 40 µM.

Magnesium is the 11th most abundant element by mass in the human body. Mg²ᶧ is essential to all living cells where it plays an important role in facilitating the processing of biological polyphosphates like ATP, DNA, RNA and enzyme functions. Mg²ᶧ is the metallic ion at the center of chlorophyll, and a common additive to fertilizers. Mg²ᶧ compounds are used as laxatives, antacids, and used to stabilize abnormal nerve excitation and blood vessel spasm i.e., eclampsia. The BioVision Magnesium Assay Kit provides a simple sensitive means of quantitating magnesium in a variety of biological samples. The kit takes advantage of the specific requirement of glycerol kinase for Mg²ᶧ. An enzyme linked reaction leads to formation of an intensely colored (λmax = 450nm) product whose formation is proportional to Mg²ᶧ concentration. The linear range of the assay is 2-15 nmoles with detection sensitivity~ 40 µM.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty