Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB102507-100TEST
10186-680EA 1081.5 USD InStock
10186-680
Zinc Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (560 nm)
Species reactivity- Mammalian
Applications- The kit can detect 0.2 µg/ml (~1-3 µM)

Zinc, a metallic chemical element, symbol Zn and atomic number 30 is chemically similar to Magnesium due to its similar size and sole oxidation state of ²ᶧ. Zinc is an essential mineral of great biological significance, because many enzymes require it as an essential cofactor. Examples of zinc’s biological roles include signal transduction, gene expression, regulation of apoptosis, synaptic plasticity and prostate gland function. BioVision’s Zinc Assay Kit is a convenient colorimetric assay in which Zinc binds to a ligand with development of absorbance at 560 nm. The assay can be used with biological samples such as serum, plasma, csf or urine with detection sensitivity 0.2 µg/ml (~1-3 µM).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty