Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB83366-100
10186-682EA 1016.07 USD InStock
10186-682
Iron Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (593 nm)
Species reactivity- Mammalian
Applications- The kit can detect 8 µM to 400 µM iron concentration in various samples.

Iron is essential to nearly all known organisms. It is generally stored in the centre of metalloproteins, in the heme complex, and in oxygen carrier proteins. Inorganic iron also contributes to redox reactions in the iron-sulfur clusters of many enzymes, such as nitrogenase and hydrogenase. BioVision's Iron Assay Kit provides a simple convenient means of measuring Ferrous and/or Ferric ion in sample. In the assay, ferric carrier protein will dissociate ferric into solution in the presence of acid buffer. After reduction to the ferrous form (Fe²ᶧ), iron reacts with Ferene S to produce a stable colored complex and give absorbance at 593 nm. A specific chelate chemical is included in the buffer to block copper ion (Cu²ᶧ) interference. The kit measures iron in the linear range of 0.4 to 20 nmol in 50 µl sample, or 8 µM to 400 µM iron concentration in various samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty