Would you like to log in again?
ApoDIRECT DNA Fragmentation Assay Kit, BioVision
Supplier: BioVision
AB66108-50
10186-686EA 863.85 USD InStock
10186-686
ApoDIRECT DNA Fragmentation Assay Kit, BioVision
Assays Cellular Assays
Detects DNA Fragmentation by FACS or FL
Detection method- Flow cytometry (Ex/Em = 488/520 nm for FITC, and 488/623 nm for PI) and fluorescence microscopy (FITC and rhodamine filters)
Species reactivity- Mammalian
Applications- The TUNEL-based assay kit provides complete components including positive and negative control cells for convenient detection of DNA fragmentation in cultured cells and tissue sections.
Internucleosomal DNA fragmentation is a hallmark of apoptosis in mammalian cells. BioVision’s ApoDIRECT In Situ DNA Fragmentation Assay Kit provides complete components including positive and negative control cells for conveniently detecting DNA fragmentation by fluorescence microscopy or flow cytometry. The TUNEL-based detection kit utilizes terminal deoxynucleotidyl transferase (TdT) to catalyze incorporation of fluorescein-12-dUTP at the free 3’-hydroxyl ends of the fragmented DNA. The fluorescein-labeled DNA can then be observed by fluorescence microscopy or analyzed by flow cytometry.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty