Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alkaline Phosphatase Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB83369-500
10186-696EA 1068.47 USD InStock
10186-696
Alkaline Phosphatase Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive Assay, HTS

Detection method- Absorbance (405 nm)
Species reactivity- Mammalian
Applications- The Kit can detect 10-250 µU ALP in samples.

Alkaline phosphatase (ALP) catalyzes the hydrolysis of phosphate esters in alkaline buffer and produces an organic radical and inorganic phosphate. Changes in alkaline phosphatase level and activity are associated with various disease states in the liver and bone. BioVision’s Alkaline Phosphatase Assay Kit is a highly sensitive, simple, direct and HTS-ready colorimetric assay designed to measure ALP activity in serum and biological samples. It contains 10 substrate tablets providing convenience for multiple usages. The kit uses p-nitrophenyl phosphate (pNPP) as a phosphatase substrate which turns yellow (λmax= 405 nm) when dephosphorylated by ALP. The Kit can detect 10-250 µU ALP in samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty