Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PicoProbe™ Phosphate Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB273281-100TEST
10186-698EA 750.77 USD InStock
10186-698
PicoProbe™ Phosphate Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive Assay

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- All
Applications- PicoProbe assay is highly sensitive with the detecting limit approximately 40 pmol/well.

Inorganic phosphate (Pi) is one of the most important ions in biological systems. It functions in a variety of roles. One of the most important roles is as a molecular switch, turning enzyme activity on and off through the mediation of the various protein kinases and phosphatases in biological systems. The Pi ion is also of great importance in mineralization processes and is a primary stimulus of algal blooms frequently found in bodies of fresh water, due to runoff from areas of high fertilizer use. The PicoProbe Phosphate Assay Kit provides an easy, quick and sensitive means of assessing phosphate over a wide range of concentrations. In the assay, inorganic phosphate will react with substrate and PicoProbe™ to generate fluorescence (Ex/Em = 535/587 nm). The kit can be used to detect Pi in a variety of samples or to monitor phosphate released by an assortment of enzymes (such as ATPases, GTPases, 5’-nucleotidase, protein phosphatases, acid and alkaline phosphatases, and phosphorylase kinase). Unlike other commercially available assays, the PicoProbe assay is not affected by the presence of glucose in samples (Note: Glucose interferes with many other commercially available assays). This PicoProbe assay is highly sensitive with the detecting limit approximately 40 pmol/well.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty