Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphate Fluorometric Assay Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB102508-100TEST
10186-700EA 489.56 USD InStock
10186-700
Phosphate Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Fluorescence (Ex/Em 535/587nm)
Species reactivity- Mammalian
Applications- Phosphate concentrations between 0.4 µM and 10 µM, with a lower detection limit of approximately 40 pmol can be directly determined.

Inorganic phosphate (Pi) is one of the most important ions in biological systems. It functions in a variety of roles. One of the most important roles is as a molecular switch, turning enzyme activity on and off through the mediation of the various protein kinases and phosphatases in biological systems. A highly sensitive assay is desired to monitor Pi in variety samples or monitor Pi changes during kinase and phosphatase reactions. The newly designed Phosphate Assay Kit provides a sensitive, easy, quick means of assessing phosphate over a wide range of concentrations. In the assay, inorganic phosphate reacted with maltose to produce glucose in the present of a special enzyme. The glucose is specifically oxidized to generate a product that reacts with the OxiRed® probe to generate fluorescence (Ex/Em=535/587nm). The kit can be used to detect Pi produced through reactions involving ATPases, GTPases, 5’-nucleotidase, protein phosphatases, acid and alkaline phosphatases, and phosphorylase, etc. from a variety of samples. Glucose background can be eliminated by doing a glucose control. Phosphate concentrations between 0.4 µM and 10 µM, with a lower detection limit of approximately 40 pmol can be directly determined.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty