Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acid Phosphatase Activity Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB83370-500
10186-702EA 360.69 USD InStock
10186-702
Acid Phosphatase Activity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Ultra-Sensitive Assay, HTS

Detection method- Fluorescence (Ex/Em 360/440)
Species reactivity- Mammalian
Applications- The detection sensitivity is ~1 µU, more sensitive than colorimetric assays. The kit is suitable for both research and drug discovery.

Acid phosphatase (AP) dephosphorylates phosphate groups from the phosphate esters in acid conditions. Different forms of acid phosphatase are found in different organs, and their serum levels are used as a diagnosis for disease in the corresponding organs. For example, elevated prostatic acid phosphatase levels may indicate the presence of prostate cancer and elevated tartrate resistant acid phosphatase levels may indicate the bone disease. BioVision’s Acid Phosphatase Fluorometric Assay Kit uses non-fluorescent Methylumbelliferyl phosphate disodium (MUP) as the substrate which has Ex/Em=360/440 nm when dephosphorylated by AP. The kit is an ultra-sensitive, simple, direct and HTS-ready fluorometric assay designed to measure AP activity in serum and other bio-samples. The detection sensitivity is ~1 µU, more sensitive than colorimetric assays. The kit is suitable for both research and drug discovery.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty