Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alkaline Phosphatase Activity Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB83371-500
10186-704EA 673.15 USD InStock
10186-704
Alkaline Phosphatase Activity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive Assay, HTS.

  • Simple procedure; takes less than 1 hour
  • Fast and convenient
  • The kit is more sensitive than colorimetric assays
  • The kit is suitable for both research and drug discovery

Detection method- Fluorescence (360/440)
Species reactivity- Mammalian
Applications- The assay designed to measure ALP activity in serum and bio-samples with detection sensitivity ~1 µU

Alkaline phosphatase (ALP) catalyzes the hydrolysis of phosphate esters in alkaline buffer and produces an organic radical and inorganic phosphate. The change in alkaline phosphatase level and activity is associated with a lot of diseases in the liver and bones. Alkaline phosphatase is also a popular enzyme conjugated to secondary antibody in ELISA. In BioVision’s Alkaline Phosphatase Fluorimetric Assay Kit, ALP cleaves the phosphate group of the non-fluorescent 4-Methylumbelliferyl phosphate disodium salt (MUP) substrate resulting in an intense fluorescent signal (Ex/Em = 360/440 nm). The kit is an ultra sensitive, simple, direct and HTS-ready assay designed to measure ALP activity in serum and bio-samples with detection sensitivity ~1 µU, more sensitive than colorimetric assays. The kit is suitable for both research and drug discovery.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty