Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia Colorimetric Assay Kit II, BioVision

Supplier: BioVision
AB102509-100TEST
10186-712EA 1188.86 USD InStock
10186-712
Ammonia Colorimetric Assay Kit II, BioVision
Assays Cellular Assays
Sensitive Assay, HTS

Detection method- Absorbance (670 nm)
Species reactivity- Mammalian
Applications- The kit can detect less than 1 nmol (~10 μM) ammonia using either a 96 well or 384 well microwell plate.

Ammonia is an important source of nitrogen for living systems. Nitrogen is required for the synthesis of amino acids, which are the building blocks of protein. Ammonia is a metabolic product which is created through amino acid deamination. It plays an important role in both normal and abnormal animal physiology such as acid/base balance. BioVision provides a rapid, simple, sensitive, and reliable assay suitable for research and high throughput assay of Ammonia. In this non-enzymatic assay, ammonia is used to form indophenol, a highly colored product easily quantifiable by colorimetry (OD670 nm) using a plate reader. The kit can detect less than 1 nmol (~10 μM) ammonia using either a 96 well or 384 well microwell plate.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty