Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Irisin Competitive ELISA Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB285295-96TEST
10186-752EA 1716.01 USD InStock
10186-752
Irisin Competitive ELISA Kit, BioVision
Assays Cellular Assays
Sensitive, Colorimetric Assay

Detection method- Absorbance (450 nm)
Species reactivity- human, mouse, rat, monkey
Application- Quantitative ELISA

BioVision’s Irisin (human) Competitive ELISA kit is to be used for the in vitro quantitative determination of human irisin in cell culture supernatants, serum and plasma. It should also work for the in vitro quantitative determination of irisin in mouse, rat and monkey biological samples. A polyclonal antibody recognizing native irisin reacts with a series of predetermined recombinant irisin standard proteins or samples under competition in the irisin-coated plate. Their relative reactivity is plotted with that of the standard proteins. Detection limit: 1 ng/ml. Note: The Limit of detection was measured by adding two standard deviations to the mean value of 50 zero standard. Assay Range: 0.001 ug/ml – 5 ug/ml. Irisin levels range in human plasma and serum from 0.2 to > 2 ug /ml.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty