Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB102510-100TEST
10186-822EA 1070.3 USD InStock
10186-822
Cobalt Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (475 nm)
Species reactivity- Mammalian
Applications- The assay is linear in the range 3 to 50 nmol (0.18-3 µg) Cobalt, with a detection limit of ~ 3 nmol (or ~15 µM, 0.18 µg).

Cobalt is a hard, lustrous, grey metal, a chemical element (symbol Co, atomic number 27 and atomic mass 58.9). Cobalt is an essential trace-element for all multicellular organisms as the active center of coenzymes called cobalamins. These include vitamin B-12 which is essential for plants and animals [1]. Cobalt is also an active nutrient for bacteria, algae, and fungi, and may be a necessary nutrient for all life. Cobalt, particularly the Co²ᶧ species, forms complexes with a large number of organic and inorganic molecules with a range of extinction coefficients in the visible range. The BioVision Cobalt Assay kit takes advantage of the reaction of Co²ᶧ with mercaptoethanol in basic media. Under the condition, Cobalt forms a complex with a strong absorbance band at 475 nm. The metal ions Fe²ᶧ, Cu²ᶧ, Ni²ᶧ, Zn²ᶧ, Mn²ᶧ, exhibit less than 10% interference at this wavelength. The kit provides a quick simple accurate method of quantitating Co²ᶧ in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty