Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyrosinase Inhibitor Screening Kit (Colorimetric), BioVision

Supplier: BioVision
AB204715-100TEST
10186-840EA 1564.98 USD InStock
10186-840
Tyrosinase Inhibitor Screening Kit (Colorimetric), BioVision
Assays Cellular Assays
Simplest High-Throughput Assay To Screen Tyrosinase Inhibitors

Detection method- Absorbance (OD 510 nm)
Applications- Screening/studying/characterizing tyrosinase inhibitors

Tyrosinase or polyphenol oxidase (EC 1.14.18.1), is an oxidoreductase that participates in the biosynthesis of melanin, a ubiquitous biological pigment found in hair, eyes, skin, etc. Inhibition of tyrosinase has been a long-time target in the skin health research, cosmetics and agricultural industries because of its role in browning reactions in skin pigmentation and during fruit harvesting and handling. Skin whitening and bleaching products utilize natural or synthetic tyrosinase inhibitors in order to lighten the skin color. Polyphenols, benzaldehyde derivatives, long-chain lipids, steroids, and natural compounds have been used as tyrosinase inhibitors. Tyrosinase catalyzes the oxidation of tyrosine, producing a chromophore that can be detected at 510 nm. In the presence of Kojic Acid, a reversible inhibitor of tyrosinase, the rate of oxidation of the substrate is decreased. The kit provides a rapid, simple, sensitive, and reliable test suitable for high-throughput screening of tyrosinase inhibitors. The assay is also adaptable to a 384-well format.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty