Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PicoProbe™ Lipolysis (3T3-L1) Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB185434-100TEST
10186-846EA 1225.11 USD InStock
10186-846
PicoProbe™ Lipolysis (3T3-L1) Fluorometric Assay Kit, BioVision
Assays Cellular Assays
The most sensitive kit on the market

Detection method- Fluorescence (Ex/Em = 535/587 nm).
Applications- - Measurement of lipolysis in 3T3-L1 cells or adipocytes
- Screening compounds that influence lipolysis, mechanistic studies, and studies on metabolic dysfunctions

Lipolysis is the hydrolysis of triglycerides within the cell into glycerol and free fatty acids. The glycerol and free fatty acids are then released into the bloodstream or culture media. Lipolysis occurs in essentially all cells, but is most abundant in white and brown adipose tissue. Deficiencies in lipolysis lead to increased intracellular lipid accumulation, resulting in abnormal cellular physiology, hyperlipidemia, and insulin resistance. Lipolysis can be induced by catecholamines and certain hormones. The kit includes the synthetic catecholamine, Isoproterenol, which activates β-adrenergic receptors. This leads to activation of adenylate cyclase, which catalyzes the conversion of ATP to cAMP. cAMP then serves as a second messenger to activate hormone-sensitive lipase, which hydrolyzes the triglycerides. This pathway can be inhibited by insulin. BioVision’s Lipolysis (3T3-L1) Fluorometric Assay kit is simple, easy-to-use & the most sensitive kit on the market. This assay measures glycerol released from 3T3-L1 cells as early as 1 hr. after induction of lipolysis. It is suitable for measuring trace amounts of glycerol from samples. In this assay, glycerol reacts with Enzyme Mix to form an intermediate that in turn reacts with Glycerol Developer & PicoProbe™ to generate the fluorescence product. The fluorescence intensity is directly proportional to the amount of glycerol. This assay kit can detect less than 20 pmol of Glycerol.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty