Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sphingomyelin Quantification Colorimetric Assay Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB287856-96TEST
10191-544EA 703.73 USD InStock
10191-544
Sphingomyelin Quantification Colorimetric Assay Kit, BioVision
Assays Cellular Assays
The most sensitive kit on the market

Detection method- Absorbance (OD 570 nm)
Applications- - Sphingomyelin quantification in various biological samples
- Analysis of cell signaling pathway

Sphingomyelin is an important structural lipid component of cell membranes and lipoproteins. Sphingomyelin has been implicated in the pathogenesis of several diseases; therefore a sensitive and reliable technique for its quantification is of considerable importance. BioVision’s Sphingomyelin Quantification Assay Kit provides a simple and sensitive method for quantifying sphingomyelin using colorimetry. In the assay, sphingomyelin is hydrolyzed into ceramide and phosphorylcholine by sphingomyelinase. Alkaline phosphatase (ALP) dephosphorylates phosphorylcholine to choline, which then reacts with Sphingomyelin Enzyme Mix to produce an intermediate. The intermediate reacts with a highly specific probe to generate color (OD 570 nm). The kit can detect sphingomyelin in various biological samples as low as 1 µM.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty