Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glucose Colorimetric/Fluorometric Assay Kit, Abcam

Supplier: Abcam
AB65333-100
10186-850EA 846.25 USD InStock
10186-850
Glucose Colorimetric/Fluorometric Assay Kit, Abcam
Assays Cellular Assays
Sensitive Assay, HTS

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The kit detects 1-10000 µM glucose samples.

Glucose (C₆H₁₂O₆; FW: 180.16) is a very important fuel source to generate universal energy molecules ATP. Glucose level is a key diagnostic parameter for many metabolic disorders. Measurement of glucose can be very important in both research and drug discovery processes. The Glucose Assay Kit provides direct measurement of glucose in various biological samples (e.g., serum, plasma, body fluid, food, growth medium, etc.). Glucose Enzyme Mix specifically oxidizes glucose to generate a product which reacts with a dye to generate color (λ = 570 nm) and fluorescence (Ex/Em = 535/587 nm). The generated color and fluorescence is proportionally to the glucose amount. The method is rapid, simple, sensitive, and suitable for high throughput. The assay is also suitable for monitoring glucose level during fermentation and glucose feeding in protein expression processes. The kit detects 1-10000 µM glucose samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty