Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Uric Acid Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB65344-100
10186-854EA 638.05 USD InStock
10186-854
Uric Acid Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian

Uric acid in serum is the end product of purine metabolism, and is cleared through the kidney by glomerular filtration. However, human often lacks the necessary enzyme called urate oxidase (Uricase), and therefore abnormal uric acid may be accumulated in blood. Recent evidences show the close association between serum urate level and cardiovascular morbidity and mortality, especially among persons at high cardiovascular risk, including those with hypertension, diabetes and congestion heart failure. BioVision’s Uric Acid Assay Kit provides a convenient means for detecting uric acid in biological samples such as serum and urine. Pretreatment of samples are not required. Uric acid level can be measured using fluorometric (at Ex/Em = 535/587 nm) or colorimetric (at λ= 570 nm) methods.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty