Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Free Fatty Acid Quantification Colorimetric/Fluorometric Kit, BioVision

Supplier: BioVision
AB65341-100
10186-862EA 1144.26 USD InStock
10186-862
Free Fatty Acid Quantification Colorimetric/Fluorometric Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Application- The kit has a detection limit 2 µM free fatty acid in variety samples.

Fatty Acids play very important roles in normal metabolism and many disease developments. They are precursors to a number of bioactive classes of compounds such as prostaglandins, leucotrienes and others, and have been implicated in diverse functions such as autism, immune system and inflammation response. BioVision’s Free Fatty Acid Quantification Kit provides a convenient, sensitive enzyme-based method for detecting the long-chain free fatty acids in various biological samples, such as serum, plasma and other body fluids, food, growth media, etc. In the assay, Fatty Acids are converted to their CoA derivatives, which are subsequently oxidized with the concomitant generation of color or fluorescence. C-8 (octanoate) and longer fatty acids can then be easily quantified by either colorimetric (spectrophotometry at λ = 570 nm) or fluorometric (at Ex/Em = 535/587 nm) methods with detection limit 2 µM free fatty acid in variety samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty