Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Choline/Acetylcholine Quantification Colorimetric/Fluorometric Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB65345-100
10186-868EA 622.04 USD InStock
10186-868
Choline/Acetylcholine Quantification Colorimetric/Fluorometric Kit, BioVision
Assays Cellular Assays
Highly Sensitive Assay

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Application- The kit can detect 10 pmol to ~5 nmol of choline or acetylcholine.

Choline and acetylcholine play important roles in many biological processes. BioVision’s Choline/Acetylcholine Quantification Kit provides a simple and sensitive means for quantifying Choline and Acetylcholine by either a colorimetric or fluorometric method. In the assay free choline is oxidized to betaine, via the intermediate betaine aldehyde. The reaction generates products which react with the Choline Probe to generate color (λ= 570 nm), and fluorescence (Ex/Em 535/587 nm). Acetylcholine can be converted to choline by adding acetylcholinesterase to the reaction. The kit can detect choline and acetylcholine (total choline – free choline) in various biological samples such as in blood, cells, culture media, fermentation media, etc. There is no need for pretreatment or purification of samples. The kit can detect 10 pmol~5 nmol of choline or acetylcholine.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty