Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glucose and Sucrose Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB65334-100
10186-870EA 911.13 USD InStock
10186-870
Glucose and Sucrose Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method-Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian

Glucose (C₆H₁₂O₆; FW: 180.16) and sucrose (C₁₂H₂₂O₁₁; FW:342.3) are the important fuel sources to generate universal energy molecule ATP. Measurement of glucose or sucrose level can be very important in both research and development process. Sucrose is a disaccharide which can be converted into one glucose and one fructose when adding Invertase. BioVision’s Glucose and Sucrose Assay Kit provides a convenient means for measuring glucose and sucrose levels from various biological samples (e.g. serum, plasma, body fluids, food, growth medium, etc.). To measure glucose level, glucose oxidase specifically oxidizes free-glucose generating a compound that reacts with the glucose probe to produce resorufin, which can be detected colorimetrically (O.D. 570 nm) or fluorometrically (Ex/Em 535/587). To measure sucrose, invertase can be added to the reaction to convert sucrose to free glucose and fructose, so total glucose level can be measured. Then the sucrose level = Total Glucose – Free Glucose.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty