Would you like to log in again?
AC Hand Generator Kit
Supplier: Avantor
WARD470199-686
470199-686KT 152.99 USD InStock
470199-686
AC Hand Generator Kit
Educational Classroom Kits and Activities
Sized for student use, this lab kit leads students in the construction of a miniature AC generator. A hand-sized model is easy for students to build and examine, with all the key components and performance of a regularly sized generator. The number of coils and speed of the magnets can be controlled and manipulated for easy student modification and multiple lab experiments.
  • Easy for students to build and examine
  • Developed by Brent Biddle
For middle school or high school.
Ordering information: This activity includes two spools of magnetic wire, 18 small magnets, six plastic spools, and instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty