Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Maltose and Glucose Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB65335-100
10186-874EA 602.6 USD InStock
10186-874
Maltose and Glucose Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Fluorescence Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Application- The assay can detect 10 pmol to 10 nmol glucose per assay.

Glucose (C₆H₁₂O₆; FW: 180.16) and Maltose (C₁₂H₂₂O₁₁; FW: 342.3) are the main fuel sources to generate the universal energy molecule ATP. Maltose is the major disaccharide that generated from hydrolysis of starch in food. Maltose contains two glucose units joined by a α-1,4-glycosidic linkage, which can be easily converted to two glucoses by α-D-glucosidase. Glucose oxidase specifically oxidizes free glucose to produce a product that interacts with the glucose probe to generate color and fluorescence. Therefore, glucose or maltose levels in various biological samples (e.g. serum, plasma, body fluids, food, growth medium, etc.) can be easily determined by either colorimetric (spectrophotometry at λ= 570 nm) or fluorometric (Ex/Em = 535/587 nm) methods. The assay can detect 10 pmol to 10 nmol glucose per assay.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty