Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB65343-100
10186-876EA 679.09 USD InStock
10186-876
Ethanol Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method-Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Sample type- Cell and Tissue culture supernatants, urine, plasma and serum, as well as many other biological fluids
Species reactivity- Mammalian
Application- The kit detects 0.1-10 ppm alcohol (~10-800 nM).

Alcohol (ethanol C₂H₅OH) is among the most widely consumed drinks. Low doses of alcohol may help circulation while heavy alcohol consumption may lead to various forms of disease. Quantitative determination of alcohol finds applications in basic research, drug discovery, clinical studies and fermentation industry processes. BioVision’s Ethanol Assay Kit provides a simple, rapid, and sensitive method for accurate quantification of ethanol concentration in a variety of biological samples such as serum, plasma, other body fluids, foods, beverages and growth media. Alcohol oxidase oxidizes ethanol to generate H₂O₂ which reacts with our probe to generate color (λmax= 570 nm) and fluorescence (Ex/Em = 535/587 nm).. The kit detects 0.1-10 ppm alcohol (~10-800 nM).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty