Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Galactose Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB83382-100
10186-878EA 556.55 USD InStock
10186-878
Galactose Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Sensitive Assay, HTS

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian

Galactose (C₆H₁₂O₆ FW: 180.16) is one the major naturally occurring sugars, involved in many biological processes. BioVision’s Galactose Assay Kit provides a simple, convenient means for direct measurement of galactose levels in various biological samples (serum, plasma, other body fluids, food, growth media, etc.). In the assay kit, Galactose is specifically oxidized generating a product that produces color (O.D 570 nm) and fluorescence (Ex/Em 535/587 nm). Liquid samples can be tested directly without purification. The assay is fast, convenient and sensitive. It can also be used as a high throughput assay.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty