Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Total Cholesterol and Cholesteryl Ester Colorimetric Assay Kit II, BioVision

Supplier: BioVision
AB102515-100TEST
10186-882EA 1235.65 USD InStock
10186-882
Total Cholesterol and Cholesteryl Ester Colorimetric Assay Kit II, BioVision
Assays Cellular Assays
Stable, Sensitive Assay

Detection method- Absorbance (450 nm)
Species reactivity- Mammalian

Cholesterol is an essential molecule in all animal life. It has been involved in both normal development and diseases. The Cholesterol/Cholesteryl Ester Quantitation Kit provides a simple method for sensitive quantification of free cholesterol, cholesteryl esters, or both using a colorimetric method. Most of the cholesterol in blood is in the form of cholesteryl ester. These esters can be hydrolyzed to free cholesterol under the appropriate conditions. In the assay, free cholesterol is oxidized by cholesterol dehydrogenase to generate NADH which reacts with a sensitive probe resulting in strong absorbance at 450 nm. The assay can detect free or total cholesterol depending upon whether esterase is utilized to hydrolyze cholesterol esters present. Cholesteryl ester can be determined by subtracting the value of free cholesterol from the total cholesterol (cholesterol plus cholesteryl esters). The probe in this kit is more stable, sensitive and specific. The assay can tolerate interferences from various samples significantly.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty