Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactose Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB83384-100
10186-884EA 591.35 USD InStock
10186-884
Lactose Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Sensitive Assay, HTS

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian

Lactose (C₁₂H₂₂O₁₁ FW: 342.3) is an important naturally occurred disaccharide, consisting of one galactose and one glucose. Milk contains ~2-8% lactose. Some people, particularly infants, lack the enzyme necessary to digest galactose leading to galactose accumulation in blood (Galactosemia) causing enlarged liver, renal failure, cataracts and brain damage. In the BioVision Lactose Assay Kit, Lactose is hydrolyzed to glucose and galactose. The galactose is subsequently oxidized generating color (O.D 570 nm) and fluorescence (Ex/Em 535/587 nm). Free galactose can be corrected by a background control in the absence of lactase. The Lactose Assay Kit provides a simple, convenient, and sensitive means for direct measurement of lactose levels in various biological samples (serum, plasma, other body fluids, food, growth media, etc.). Pretreatment of samples is not required. The kit can be used as a high throughput assay.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty