Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Creatinine Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB65340-100
10186-886EA 857.26 USD InStock
10186-886
Creatinine Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian

Creatinine is a breakdown product of creatine phosphate. Creatinine is produced and excreted at a constant rate, and blood creatinine is used to determine glomerular filtration rate (GFR), a measure of kidney function. Blood creatinine levels increase only in cases of significant (>75%) damage to nephrons. Creatinine clearance is frequently used as a means of standardizing excretion of other compounds such as isoprostanes. BioVision’s Creatinine Assay Kit provides an accurate, convenient measure of creatine concentration in biological fluids such as serum, urine or CSF. In the assay, creatinine is converted to creatine by creatininase, creatine is converted to sarcosine, which is specifically oxidized to produce a product which reacts with a probe to generate red color (λmax = 570 nm) and fluorescence (Ex/Em = 538/587 nm). Unlike the picric acid assay, this kit is suitable for serum/plasma creatinine determinations, as well as for urine and other biological samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty