Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sucrose Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB83387-100
10186-888EA 543.47 USD InStock
10186-888
Sucrose Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
No interference from Lactose & Maltose

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The method can detect 0.0002-10 mM sucrose concentrations.

Sucrose (C₁₂H₂₂O₁₁; FW:342.3), also called table sugar, is one of the most important fuel sources to generate the universal energy molecule ATP. Sucrose is a disaccharide which can be converted into one glucose and one fructose. BioVision’s Sucrose Assay Kit provides a convenient means for specifically measuring sucrose levels from various biological samples (e.g. serum, plasma, body fluids, food, growth medium, etc.). Sucrose is converted to glucose and fructose by invertase. The generated glucose is then specifically oxidized to generate a product which reacts with a probe to generate color (λ= 570 nm) and fluorescence (Ex/Em = 535/587 nm). Use of proper controls allows for correcting free glucose background. Other disaccharides such as lactose and maltose do not interfere with the assay. The method can detect 0.0002-10 mM sucrose concentrations.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty