Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactate Colorimetric Assay Kit II, BioVision

Supplier: BioVision
AB65331-100
10186-890EA 1147.97 USD InStock
10186-890
Lactate Colorimetric Assay Kit II, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Application- Kit detects 0.02 mM – 10 mM lactate in various samples.

Lactate (CH₃CH(OH)COO-) plays important roles in many biological processes. Abnormally high concentrations of lactate have been related to disease states such as diabetes and lactic acidosis, etc. L(+)-Lactate is the major lactate stereoisomer formed in human intermediary metabolism and is present in blood. D(-)-Lactate is also present but only at about 1-5% of the concentration of L(+)-Lactate. In the Lactate Assay Kit, lactate is oxidized by lactate dehydrogenase to generate a product which interacts with a probe to produce a color (λmax = 450 nm). The kit detect L(+)-Lactate in biological samples such as in serum or plasma, cells, culture and fermentation media. There is no need for pretreatment or purification of samples. It detects 0.02 mM – 10 mM lactate in various samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty