Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Maltose Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB83388-100
10186-892EA 558.41 USD InStock
10186-892
Maltose Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian

Maltose (C₁₂H₂₂O₁₁; FW: 342.3), one of the main fuel sources to generate the universal energy molecule ATP, is the major disaccharide that is generated from hydrolysis of starch in food. Maltose contains two glucose units joined by a α-1,4-glycosidic linkage, which can be easily converted to two glucoses by α-D-glucosidase. The generated glucose can be specifically oxidized to produce a product that interacts with the probe to generate color and fluorescence. Thus, maltose can be determined by either colorimetric (spectrophotometry at λ = 570 nm) or fluorometric (Ex/Em = 535/587 nm) methods. The kit provides a fast, easy and sensitive method for quantifying maltose in various biological samples (e.g. serum, plasma, body fluids, food, growth medium, etc.).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty