Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glutamate Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB83389-100
10186-894EA 980.86 USD InStock
10186-894
Glutamate Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Species reactivity- All

Glutamate, one of the two acidic proteinogenic amino acids, is also a key molecule in cellular metabolism. In humans, glutamate plays an important role both in amino acid degradation and disposal of excess or waste nitrogen. Glutamate is the most abundant swift excitatory neurotransmitter in the mammalian nervous system. It is believed to be involved in learning and memory and has appeared to be involved in diseases like amyotrophic lateral sclerosis, lathyrism, autism, some forms of mental retardation and Alzheimer's disease. Glutamic acid is also present in a wide variety of foods, and has been used as a flavor enhancer in food industry. BioVision’s Glutamate Assay Kit provides a sensitive detection method of the glutamate in a variety of samples. The glutamate Enzyme Mix recognizes glutamate as a specific substrate leading to proportional color development. The amount of glutamate can therefore be easily quantified by colorimetric (spectrophotometry at λ = 450 nm) method.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty