Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Hydroxybutyrate (β-HB) Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB83390-100
10186-900EA 643.81 USD InStock
10186-900
β-Hydroxybutyrate (β-HB) Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Application- The assay is linear for 1-20 nmol β-HB in up to 100 µl samples or 0.01-0.2 mM of β-HB samples.

Diabetic ketoacidosis occurs when circulating insulin levels drop to very low levels, shutting off the supply of glucose to the body. The physiological response is for the liver to produce ketone bodies (acetoacetate, acetone, and primarily β-hydroxybutyrate) from the acetyl CoA produced from fatty acid oxidation. The very high rate of ketone body production outstrips the body’s ability to utilize them as an energy source and the blood concentration builds up. As rather strong acids, they result in a significant drop in blood pH. BioVision’s β-HB Assay kit utilizes β-HB Dehydrogenase to generate a product which reacts with our colorimetric probe with an absorbance band at 450 nm. The kit is an easy and accurate assay to measure β-HB levels in biological samples. The assay is linear for 1-20 nmol β-HB in up to 100 µl samples or 0.01-0.2 mM of β-HB samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty