Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fumarate Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB102516-100TEST
10186-902EA 1007.21 USD InStock
10186-902
Fumarate Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Application- Kit can detect as low as 1 nmol of fumarate per well (20 µM).

Fumarate (HO₂CCH=CHCO₂H-) is an intermediate in the Kreb’s cycle used by cells to metabolize food to form ATP. In the mammalian liver, Fumarate is also a product of the Urea cycle where its release in the cytosol leads to its conversion into malate and subsequently oxaloacetate while generating NADH in the cytosol. The human skin naturally produces fumaric acid when exposed to sunlight. In fact, fumaric acid esters have been used to treat psoriasis, possibly due to an impaired production of fumaric acid in the skin. Fumaric acid has also been used in beverages, baking powders and candy. BioVision’s Fumarate Assay Kit provides a convenient tool for sensitive detection of the fumarate in a variety of samples. The fumarate Enzyme Mix recognizes fumarate as a specific substrate leading to proportional color development. The amount of fumarate can therefore be easily quantified using a colorimetric assay (λ = 450 nm). It can detect as low as 1 nmol of fumarate per well (20 µM).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty