Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Creatine Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB65339-100
10186-904EA 1098.15 USD InStock
10186-904
Creatine Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications-The kit can detect 0.001 – 10 mM Creatine.

Creatine is an endogenous compound whose function is to maintain a high ATP/ADP ratio, by way of its phosphorylated form and creatine kinase. Creatine supplementation has been used in the treatment of muscular, neurological and neurodegenerative diseases, as well as a sport performance enhancer. Detection of creatine level has wide applications in research and development. BioVision’s Creatine Assay Kit provides an accurate, convenient measure of creatine in a variety of biological samples. In the assay, creatine is enzymatically converted to sarcosine which is then specifically oxidized to generate a product that converts a colorless probe to an intensely red color (λmax = 570 nm), and highly fluorescent (Ex/Em = 538/587 nm) product. Creatine is therefore easily detected by either colorimetric or fluorometric methods. Detection range 0.001 – 10 mM Creatine.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty