Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sarcosine Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB65338-100
10186-906EA 518.06 USD InStock
10186-906
Sarcosine Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications-The kit can detect Sarcosine in the detection range of 1 – 10000 µM.

Sarcosine, a natural amino acid, plays important roles as intermediate in the metabolism of choline, methionine, glycine, glutathione, creatine, purine and serine, etc. Detection of sarcosine level has wide applications in research and development. BioVision’s Sarcosine Assay Kit provides an accurate, convenient measure of sarcosine in variety biological samples. In the assay, sarcosine is specifically oxidized to generate a product that converts a colorless probe to a product with intense red color (λmax = 570 nm) and which is also highly fluorescent (Ex/Em = 538/587 nm). Sarcosine is therefore easily detected by either colorimetric or fluorometric methods with detection range 1 – 10000 µM.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty