Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycerol-3-Phosphate (G3P) Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB174094-100TEST
10186-914EA 926.96 USD InStock
10186-914
Glycerol-3-Phosphate (G3P) Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Applications:
-Measurement of Glycerol-3-Phosphate in various tissues/cells
-Analysis of carbohydrate and lipid metabolism and cell signaling
-Drug screening

Glycerol-3-phosphate (G3P) is an important intermediate for all living organisms. Glycerol-3-Phosphate is produced either by glycerol via glycerol kinase or by dihydroxyacetone phosphate through glycerol-3-phosphate dehydrogenase. In response to cellular signals, glycerol-3-phosphate can be utilized in multiple pathways: it can be further converted into glyceraldehyde-3-phosphate and enter glycolysis or rapidly generate NAD+ in brain or muscle tissues through the G3P shuttle or enter the lipid biosynthetic pathway. Recent studies have found that glycerol-3-phosphate is a novel regulator and plays a fundamental defense role in plant pathogenesis. BioVision’s Glycerol-3-Phosphate Assay kit is a sensitive, fast and easy-to-use kit. In this assay, G3P is oxidized by G3P Enzyme Mix to form an intermediate, which reduces a nearly colorless probe to a colored product with strong absorbance at 450 nm. This assay kit can detect G3P less than 2 nmol/well and can be used for a variety of sample types.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty