Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Carnitine Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB83392-100
10186-916EA 648.22 USD InStock
10186-916
L-Carnitine Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The detection sensitivity is ~1 µM for the fluorometric assay and ~10 µM for the colorimetric assay.

Carnitine is a quaternary ammonium compound biosynthesized from the amino acids lysine and methionine. It is required for transport of fatty acids into the mitochondrial matrix via the carnitine/acylcarnitine shuttle where β-oxidation occurs, acetate is generated and the acetate utilized in the TCA cycle for the generation of energy. L-Carnitine is often sold as a nutritional supplement. Carnitine exists in two stereoisomers. Only L-carnitine is biologically active. BioVision's L-Carnitine Assay Kit is a simple convenient means of measuring free L-Carnitine in biological samples such as serum. The assay transfers an acetyl group from CoA to carnitine and the free CoA formed is further processed with subsequent oxidation of the Oxi-Red probe to give fluorescence (Ex/Em 535 nm 587 nm) and absorbance (570 nm). The normal range for serum L-Carnitine is ~20-100 µM. The detection sensitivity is ~1 µM for the fluorometric assay and ~10 µM for the colorimetric assay.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty