Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Mannitol Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB155890-100TEST
10186-920EA 1093.84 USD InStock
10186-920
D-Mannitol Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Application- The kit can detect Mannitol concentration less than 1 nmol (< 10 µM).

D-Mannitol is one of the most abundant sugar alcohols in nature. It can be produced in the biosphere by a range of organisms such as bacteria, yeast, fungi and almost all plants. Mannitol functions as a carbohydrate storage depot, as a scavenger for reactive oxygen species and as an osmoprotectant. Recent studies have shown that increased mannitol content in plants protects them against fungal infections and improves resistance to drought and high salinity. The mannitol pathway reversibly converts fructose to mannitol and serves as the main carbohydrate cycle for fungi survival. Measurement of mannitol level is important for evaluating mannitol metabolic pathways and commercially, in developing fungi and drought resistant plants. In BioVision’s D-Mannitol Assay Kit, D-mannitol is converted to D-fructose by mannitol dehydrogenase in the presence of NAD to form NADH, which reduces a colorless probe to a chromogen with strong absorbance at 450 nm. L-Arabitol, another sugar alcohol, also acts as substrate for mannitol dehydrogenase & undergoes similar reaction; therefore, this kit can also be used to measure L-arabitol. The D-mannitol assay kit is fast (~15 minutes), sensitive & easy to use. This assay kit can detect mannitol level less than 1 nmol/reaction (< 10 µM) & can be used for a variety of sample types.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty