Would you like to log in again?
Total Carbohydrate Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB155891-100TEST
10186-922EA 1056.58 USD InStock
10186-922
Total Carbohydrate Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Robust, Sensitive Assay
Detection method- Absorbance (490 nm)
Can detect most forms of carbohydrates, including simple & complex saccharides, glycans, glycoproteins & glycolipids
Carbohydrates play important structural as well as chemical roles in all living systems. Detection of total carbohydrates, therefore, has wide applications. BioVision’s Total Carbohydrate Assay is a simple, sensitive and robust method of detecting virtually all carbohydrates. The assay is based on the phenol-sulfuric acid method. In Total Carbohydrate Assay, polysaccharides (mono, di, tri, etc.) and their derivatives, in the presence of sulfuric acid, are hydrolyzed to monomers and converted to furfural or hydroxyfurfural, which react with the Developer to form a chromogen that can be quantified by measuring the absorbance at 490 nm. The Total Carbohydrate Assay can detect most forms of carbohydrates, including simple and complex saccharides, glycans, glycoproteins and glycolipids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty