Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycogen Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB65620-100
10186-924EA 1052.37 USD InStock
10186-924
Glycogen Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The assay can detect glycogen 0.0004 to 2 mg/ml.

Glycogen is the primary short term energy storage molecule in animals, synthesized primarily in the liver and muscle. Glycogen is a branched glucose polymer, in α-1,4 linkage, with branching via α-1,6 linkage. Abnormal ability to utilize glycogen is found in diabetes and in several genetic glycogen storage diseases. The BioVision Kit is an easy and accurate assay to measure glycogen levels in biological samples. In the assay, glucoamylase hydrolyzes the glycogen to glucose which is then specifically oxidized to produce a product that reacts with OxiRed probe to generate color (570 nm) and fluorescence (Ex 535/Em 587). The assay can detect glycogen 0.0004 to 2 mg/ml.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty