Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycogen Colorimetric Assay Kit II, BioVision

Supplier: BioVision
AB169558-100TEST
10191-556EA 911.64 USD InStock
10191-556
Glycogen Colorimetric Assay Kit II, BioVision
Assays Cellular Assays
For samples having reducing substances

Detection method- Absorbance (450 nm)
Applications- The assay can detect less than 4 µg/ml of Glycogen in various tissues. Analysis of metabolism and cell signaling.

Glycogen serves as the main carbohydrate storage in animals and can be converted to glucose readily. It is primarily found in the liver and muscle tissues. Glycogen is a branched biopolymer comprising of α-1,4 linkage with α-1,6 linkages occurring every 8-10 glucose units along the backbone. Abnormal ability to utilize glycogen is found in diabetes and in several genetic glycogen storage diseases. Biovision’s Glycogen Assay kit II provides a simple, fast and robust way to measure Glycogen levels in various biological samples. This assay is suitable for measuring Glycogen levels in samples that contain reducing substances, which may interfere with the oxidase-based assays. In this assay, Glycogen is hydrolyzed into glucose, which is oxidized to form an intermediate that reduces a colorless Probe to a colored product with strong absorbance at 450 nm. This high-throughput suitable assay kit can detect less than 4 µg/ml of Glycogen in samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty