Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Succinate (Succinic Acid) Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB204718-100TEST
10191-558EA 1388.92 USD InStock
10191-558
Succinate (Succinic Acid) Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Only kit on the market to measure succinate (succinic acid)

Detection method- Absorbance (450 nm)
Applications- Measurement of succinate or succinic acid in various samples
- Analysis of citric acid cycle in various tissues/cells

Succinic acid (C4H6O4) is distributed in all plants and animal tissues and was first obtained from amber. Due to its low toxicity, it is widely used in agriculture, food and pharmaceutical industry. Succinate (C4H4O4) is the salt or ester of succinic acid. It is one of the most active components of cellular respiratory and intracellular energy generation. Measurement of succinate or succinic acid level is a key to analysis of the citric acid cycle. BioVision’s Succinate (Succinic Acid) Assay kit is a sensitive, fast and easy-to-use kit. In this assay, Succinate is utilized by Succinyl-CoA Synthetase to form an intermediate, which undergoes a series of reactions & reduces a colorless probe to a colored product with strong absorbance at 450 nm. This assay kit can detect less than 40 µM of succinate or succinic acid in a variety of sample types.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty