Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetoacetate Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB180875-100TEST
10186-926EA 1306.17 USD InStock
10186-926
Acetoacetate Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Only kit on the market to measure acetoacetate

Detection method- Absorbance (550 nm)
Applications: - Measurement of acetoacetate in various biological samples
Analysis of acetoacetate in pathological conditions such as Type I diabetes, alcoholism, etc.

Acetoacetate (AcAc), a β-ketoacid, is one of the three ketone bodies and is formed via condensation of two molecules of acetyl-CoA in liver mitochondria. AcAc can be enzymatically reduced to 3-β-hydroxybutyrate (β-HB), or decarboxylated producing acetone (CH3)2CO). Ketone bodies (βHB: 78%; AcAc: 20% & CH3)2CO: 2%) are mainly used as an alternative energy source when glucose cannot be delivered to the system. Excessive concentration of ketone bodies (ketoacidosis) is observed in patients with Type I diabetes, severe starvation or alcoholism. Traditionally, AcAc levels have been qualitatively detected using dipsticks that use sodium nitroferricyanide as a chromophore. BioVision’s Acetoacetate Assay Kit has adapted that principle with a modification that provides a sensitive method to quantitate endogenous levels of AcAc in human blood, and urine. In this non-enzymatic assay, AcAc reacts with a substrate to generate a colored product that can be measured at 550 nm. The reaction is specific for AcAc and does not detect 3-β-hydroxybutyrate. The assay kit can detect samples containing acetoacetate as low as 25 µM.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty