Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alanine Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB83394-100
10186-930EA 774.96 USD InStock
10186-930
Alanine Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The assay can detect serum concentration: ~24-76 µg/ml (~3-9 nmol/10 µl).

Alanine is the 2nd most abundant of the 20 proteinogenic amino acids. Nonessential, being available from dietary sources, it plays a key role in the glucose-alanine cycle between tissues and liver. In muscle and other tissues that degrade amino acids, amino groups are pooled as glutamate by transamination. Glutamate then transfers the amino group to pyruvate via alanine aminotransferase, forming alanine and α-ketoglutarate. The alanine is passed into the blood and transported to the liver. A reverse of the alanine aminotransferase reaction takes place in liver. Pyruvate can be used in gluconeogenesis, to form glucose which may return to other tissues through the circulatory system. There appears to be a correlation between alanine levels and and higher blood pressure, energy intake, cholesterol levels, and body mass index. BioVision’s Alanine Assay Kit provides a sensitive detection method of alanine. In the kit, alanine is converted to pyruvate which is specifically detected leading to proportional color (λ=570nm: 0-10 nmol) or fluorescence (Ex/Em 535/587nm: 0-1 nmol) development. Serum concentration: ~24-76 µg/ml (~3-9 nmol/10 µl).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty